missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-3-chloro-1-phenyl-1-propanol, 97%, Thermo Scientific™

Chemical Identifiers

CAS 100306-33-0
Molecular Formula C9H11ClO
Molecular Weight (g/mol) 170.64
InChI Key JZFUHAGLMZWKTF-SECBINFHSA-N
Synonym r-3-chloro-1-phenylpropan-1-ol, r-+-3-chloro-1-phenyl-1-propanol, r-+-3-chloro-1-phenylpropanol, 1r-3-chloro-1-phenylpropan-1-ol, 1r-3-chloro-1-phenyl-propan-1-ol, r-3-chloro-1-phenyl-1-propanol, r-3-chloro-1-phenyl-propan-1-ol, r-3-chloro-1-phenylpropanol, pubchem5738
PubChem CID 642409
IUPAC Name (1R)-3-chloro-1-phenylpropan-1-ol
SMILES C1=CC=C(C=C1)C(CCCl)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC432040010
SDS View Documents
Thermo Scientific Chemicals
432040010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC432040050
View Documents
Thermo Scientific Chemicals
432040050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

100306-33-0
170.64
r-3-chloro-1-phenylpropan-1-ol, r-+-3-chloro-1-phenyl-1-propanol, r-+-3-chloro-1-phenylpropanol, 1r-3-chloro-1-phenylpropan-1-ol, 1r-3-chloro-1-phenyl-propan-1-ol, r-3-chloro-1-phenyl-1-propanol, r-3-chloro-1-phenyl-propan-1-ol, r-3-chloro-1-phenylpropanol, pubchem5738
(1R)-3-chloro-1-phenylpropan-1-ol
C9H11ClO
JZFUHAGLMZWKTF-SECBINFHSA-N
642409
C1=CC=C(C=C1)C(CCCl)O

Specifications

100306-33-0
100.0
96% min. (GC)
1g
r-3-chloro-1-phenylpropan-1-ol, r-+-3-chloro-1-phenyl-1-propanol, r-+-3-chloro-1-phenylpropanol, 1r-3-chloro-1-phenylpropan-1-ol, 1r-3-chloro-1-phenyl-propan-1-ol, r-3-chloro-1-phenyl-1-propanol, r-3-chloro-1-phenyl-propan-1-ol, r-3-chloro-1-phenylpropanol, pubchem5738
C1=CC=C(C=C1)C(CCCl)O
170.64
170.64
(R)-(+)-3-chloro-1-phenyl-1-propanol
96.0
Authentic
C9H11ClO
+23° to +29° (20°C, 589nm) (c=1 in Chloroform)
JZFUHAGLMZWKTF-SECBINFHSA-N
(1R)-3-chloro-1-phenylpropan-1-ol
642409
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only