accessibility menu, dialog, popup

UserName

ClOH

CAS RN 100306-33-0

IUPAC Name: 3-chloro-1-phenylpropan-1-ol
Synonyms: 3-chloro-1-phenylpropan-1-ol
Molecular Weight (g/mol): 170.64
Molecular Formula: C9H11ClO
InChi Key: JZFUHAGLMZWKTF-UHFFFAOYNA-N
SMILES: OC(CCCl)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)