missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-3-chloro-1-phenyl-1-propanol, 97%

Catalog No. AC432040010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC432040010 1 g
AC432040050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC432040010 Supplier Thermo Scientific Chemicals Supplier No. 432040010

CAS: 100306-33-0 | C9H11ClO | 170.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 100306-33-0
Molecular Formula C9H11ClO
Molecular Weight (g/mol) 170.64
InChI Key JZFUHAGLMZWKTF-SECBINFHSA-N
Synonym r-3-chloro-1-phenylpropan-1-ol, r-+-3-chloro-1-phenyl-1-propanol, r-+-3-chloro-1-phenylpropanol, 1r-3-chloro-1-phenylpropan-1-ol, 1r-3-chloro-1-phenyl-propan-1-ol, r-3-chloro-1-phenyl-1-propanol, r-3-chloro-1-phenyl-propan-1-ol, r-3-chloro-1-phenylpropanol, pubchem5738
PubChem CID 642409
IUPAC Name (1R)-3-chloro-1-phenylpropan-1-ol
SMILES C1=CC=C(C=C1)C(CCCl)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 59.5°C to 59.8°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 1 g
Specific Rotation +23° to +29° (20°C, 589nm) (c=1 in Chloroform)
Formula Weight 170.64
Percent Purity 97%
Physical Form Powder
Chemical Name or Material (R)-(+)-3-chloro-1-phenyl-1-propanol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

RUO – Research Use Only