missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-3-Chloro-1,2-propanediol 96.0+%, TCI America™

Catalog No. p-7132467
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M096710G 10 g
M096725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M096710G Supplier TCI America Supplier No. M096710G
Only null left

Chemical Identifiers

CAS 57090-45-6
Molecular Formula C3H7ClO2
Molecular Weight (g/mol) 110.537
MDL Number MFCD00135169
InChI Key SSZWWUDQMAHNAQ-VKHMYHEASA-N
Synonym r---3-chloro-1,2-propanediol, r-3-chloro-1,2-propanediol, 2r-3-chloropropane-1,2-diol, r-3-chloropropane-1,2-diol, alpha-chlorohydrin,-, unii-64ymo02kae, ccris 7387, 1,2-propanediol, 3-chloro-, r, 64ymo02kae, 1,2-propanediol, 3-chloro-, 2r
PubChem CID 148793
ChEBI CHEBI:18663
IUPAC Name (2R)-3-chloropropane-1,2-diol
SMILES C(C(CCl)O)O

Specifications

Boiling Point 95°C
Quantity 10 g
UN Number 2689
Formula Weight 110.54
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-(-)-3-Chloro-1,2-propanediol
EINECSNumber (2)-2441
TSCA No