accessibility menu, dialog, popup

UserName

OH(R)HOCl

CAS RN 57090-45-6

IUPAC Name: (2R)-3-chloropropane-1,2-diol
Synonyms: (2R)-3-chloropropane-1,2-diol
Molecular Weight (g/mol): 110.54
Molecular Formula: C3H7ClO2
InChi Key: SSZWWUDQMAHNAQ-VKHMYHEASA-N
SMILES: OC[C@@H](O)CCl
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)