Learn More
(R)-(-)-3-Chloro-1,2-propanediol, 97%, ee 98%
CAS: 57090-45-6 | C3H7ClO2 | 110.537 g/mol
Supplier: Thermo Scientific Chemicals L1855514
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| (R)-(-)-3-Chloro-1,2-propanediol | |
| 1.322 | |
| 137°C (279°F) | |
| C3H7ClO2 | |
| MFCD00135169 | |
| UN2689 | |
| r---3-chloro-1,2-propanediol, r-3-chloro-1,2-propanediol, 2r-3-chloropropane-1,2-diol, r-3-chloropropane-1,2-diol, alpha-chlorohydrin,-, unii-64ymo02kae, ccris 7387, 1,2-propanediol, 3-chloro-, r, 64ymo02kae, 1,2-propanediol, 3-chloro-, 2r | |
| −0.9° (Neat) | |
| (2R)-3-chloropropane-1,2-diol | |
| 148793 | |
| 110.54 |
| 57090-45-6 | |
| 213°C | |
| ee 98% | |
| 1.48 | |
| 25 g | |
| 2202928 | |
| SSZWWUDQMAHNAQ-VKHMYHEASA-N | |
| C(C(CCl)O)O | |
| 110.537 | |
| CHEBI:18663 | |
| 97% |
Chemical Identifiers
| 57090-45-6 | |
| 110.537 | |
| SSZWWUDQMAHNAQ-VKHMYHEASA-N | |
| 148793 | |
| (2R)-3-chloropropane-1,2-diol |
| C3H7ClO2 | |
| MFCD00135169 | |
| r---3-chloro-1,2-propanediol, r-3-chloro-1,2-propanediol, 2r-3-chloropropane-1,2-diol, r-3-chloropropane-1,2-diol, alpha-chlorohydrin,-, unii-64ymo02kae, ccris 7387, 1,2-propanediol, 3-chloro-, r, 64ymo02kae, 1,2-propanediol, 3-chloro-, 2r | |
| CHEBI:18663 | |
| C(C(CCl)O)O |
Safety and Handling
GHS H Statement
H301-H331-H360-H341-H318-H312-H315-H335
Toxic if swallowed.
Toxic if inhaled.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Causes serious eye damage.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P312-P330-P332+P313-P362-P501c
H301-H312-H315-H318-H330-H335-H341-H360
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: GLYCEROL ALPHA-MONOCHLOROHYDRIN
EINECSNumber : 435-560-9
RTECSNumber : TY4025000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only