missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-3-Chloro-1,2-propanediol, 97%, ee 98%

Catalog No. AAL1855506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1855506 5 g
AAL1855514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1855506 Supplier Thermo Scientific Chemicals Supplier No. L1855506

CAS: 57090-45-6 | C3H7ClO2 | 110.537 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 57090-45-6
Molecular Formula C3H7ClO2
Molecular Weight (g/mol) 110.537
MDL Number MFCD00135169
InChI Key SSZWWUDQMAHNAQ-VKHMYHEASA-N
Synonym r---3-chloro-1,2-propanediol, r-3-chloro-1,2-propanediol, 2r-3-chloropropane-1,2-diol, r-3-chloropropane-1,2-diol, alpha-chlorohydrin,-, unii-64ymo02kae, ccris 7387, 1,2-propanediol, 3-chloro-, r, 64ymo02kae, 1,2-propanediol, 3-chloro-, 2r
PubChem CID 148793
ChEBI CHEBI:18663
IUPAC Name (2R)-3-chloropropane-1,2-diol
SMILES C(C(CCl)O)O

Specifications

Density 1.322
Boiling Point 213°C
Flash Point 137°C (279°F)
Refractive Index 1.48
Quantity 5 g
UN Number UN2689
Beilstein 2202928
Optical Rotation −0.9° (Neat)
Formula Weight 110.54
Percent Purity 97%
Assay ee 98%
Chemical Name or Material (R)-(-)-3-Chloro-1,2-propanediol
Show More Show Less
Hazard Category H301-H312-H315-H318-H330-H335-H341-H360
Hazard Statement GHS H Statement
H301-H331-H360-H341-H318-H312-H315-H335
Toxic if swallowed.
Toxic if inhaled.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Causes serious eye damage.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: GLYCEROL ALPHA-MONOCHLOROHYDRIN
EINECSNumber 435-560-9
RTECSNumber TY4025000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only