missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-Bromo-alpha-methylbenzyl alcohol, 95%, 98% ee

Catalog No. p-3211067
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC376490010 1 g Glass bottle
AC376490050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC376490010 Supplier Thermo Scientific Chemicals Supplier No. 376490010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 134615-24-0 | C8H9BrO | 201.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 134615-24-0
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.06
MDL Number MFCD06658929
InChI Key ULMJQMDYAOJNCC-ZCFIWIBFSA-N
Synonym r-1-3-bromophenyl ethanol, 1r-1-3-bromophenyl ethanol, 1r-1-3-bromophenyl ethan-1-ol, pubchem15565, r-1-m-bromophenyl ethanol, r-1-3-bromo-phenyl-ethanol, r-3-bromo-alpha-methylbenzyl alcohol, benzenemethanol, 3-bromo-a-methyl-, ar
PubChem CID 2734889
IUPAC Name (1R)-1-(3-bromophenyl)ethanol
SMILES CC(C1=CC(=CC=C1)Br)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless
Enantiomeric Excess 97.5% min. (GC)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH(OH)C6H4Br
Quantity 1 g
Formula Weight 201.06
Percent Purity 95%,98% ee
Physical Form Liquid
Chemical Name or Material (R)-3-Bromo-α-methylbenzyl alcohol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only