missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-3-(Boc-amino)-3-phenylpropionic acid, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5198403
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| (R)-3-(Boc-amino)-3-phenylpropionic acid | |
| C14H19NO4 | |
| 1 g | |
| r-n-boc-3-amino-3-phenylpropanoic acid, r-3-tert-butoxycarbonyl amino-3-phenylpropanoic acid, boc-r-3-amino-3-phenylpropionic acid, boc-beta-phe-oh, r-3-boc-amino-3-phenylpropionic acid, r-3-boc-amino-3-phenylpropanoic acid, boc-r-phenylalanine, 3r-3-tert-butoxycarbonyl amino-3-phenylpropanoic acid, r-3-tert-butoxycarbonylamino-3-phenyl-propionic acid | |
| CC(C)(C)OC(=O)NC(CC(=O)O)C1=CC=CC=C1 | |
| 265.309 | |
| 265.31 |
| 161024-80-2 | |
| MFCD01320859 | |
| 5404389 | |
| JTNQFJPZRTURSI-LLVKDONJSA-N | |
| (3R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoic acid | |
| 7009108 | |
| 95% |
Chemical Identifiers
| 161024-80-2 | |
| 265.309 | |
| JTNQFJPZRTURSI-LLVKDONJSA-N | |
| 7009108 | |
| CC(C)(C)OC(=O)NC(CC(=O)O)C1=CC=CC=C1 |
| C14H19NO4 | |
| MFCD01320859 | |
| r-n-boc-3-amino-3-phenylpropanoic acid, r-3-tert-butoxycarbonyl amino-3-phenylpropanoic acid, boc-r-3-amino-3-phenylpropionic acid, boc-beta-phe-oh, r-3-boc-amino-3-phenylpropionic acid, r-3-boc-amino-3-phenylpropanoic acid, boc-r-phenylalanine, 3r-3-tert-butoxycarbonyl amino-3-phenylpropanoic acid, r-3-tert-butoxycarbonylamino-3-phenyl-propionic acid | |
| (3R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoic acid |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only