missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-(Boc-amino)-3-phenylpropionic acid, 95%, Thermo Scientific Chemicals

Catalog No. p-7034313
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH51984MD 250 mg
AAH5198403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH51984MD Supplier Thermo Scientific Chemicals Supplier No. H51984MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 161024-80-2 | Molecular Formula: C14H19NO4 | Molecular Weight: 265.309 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 161024-80-2
Molecular Formula C14H19NO4
Molecular Weight (g/mol) 265.309
MDL Number MFCD01320859
InChI Key JTNQFJPZRTURSI-LLVKDONJSA-N
Synonym r-n-boc-3-amino-3-phenylpropanoic acid, r-3-tert-butoxycarbonyl amino-3-phenylpropanoic acid, boc-r-3-amino-3-phenylpropionic acid, boc-beta-phe-oh, r-3-boc-amino-3-phenylpropionic acid, r-3-boc-amino-3-phenylpropanoic acid, boc-r-phenylalanine, 3r-3-tert-butoxycarbonyl amino-3-phenylpropanoic acid, r-3-tert-butoxycarbonylamino-3-phenyl-propionic acid
PubChem CID 7009108
IUPAC Name (3R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)O)C1=CC=CC=C1

Specifications

Quantity 250 mg
Beilstein 5404389
Formula Weight 265.31
Percent Purity 95%
Chemical Name or Material (R)-3-(Boc-amino)-3-phenylpropionic acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only