missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-Amino-3-phenylpropionic acid, 95%

CAS: 83649-48-3 | C9H12ClNO2 | 201.65 g/mol

Supplier:  Thermo Scientific Chemicals H52034MD

 View more versions of this product

Catalog No. AAH52034MD


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(R)-3-Amino-3-phenylpropionic acid
C9H12ClNO2
r-3-amino-3-phenylpropanoic acid hydrochloride, r-3-amino-3-phenylpropionic acid hydrochloride, 3r-3-amino-3-phenylpropanoic acid hydrochloride, l-beta-homophenylglycine hydrochloride, r-3-amino-3-phenylpropionic acid hcl, pubchem5925, d-beta-phenylalanine hcl, r-beta-homophenylglycine hydrochloride, 3r-3-amino-3-phenylpropanoic acid, chloride, r-3-amino-3-phenyl-propionic acid hcl salt
C1=CC=C(C=C1)C(CC(=O)O)N.Cl
250 mg
2734697
Air Sensitive
83649-48-3
MFCD01076237
ABEBCTCOPRULFS-DDWIOCJRSA-N
(3R)-3-amino-3-phenylpropanoic acid;hydrochloride
201.65
165.19
95%

Chemical Identifiers

83649-48-3
201.65
ABEBCTCOPRULFS-DDWIOCJRSA-N
2734697
C1=CC=C(C=C1)C(CC(=O)O)N.Cl
C9H12ClNO2
MFCD01076237
r-3-amino-3-phenylpropanoic acid hydrochloride, r-3-amino-3-phenylpropionic acid hydrochloride, 3r-3-amino-3-phenylpropanoic acid hydrochloride, l-beta-homophenylglycine hydrochloride, r-3-amino-3-phenylpropionic acid hcl, pubchem5925, d-beta-phenylalanine hcl, r-beta-homophenylglycine hydrochloride, 3r-3-amino-3-phenylpropanoic acid, chloride, r-3-amino-3-phenyl-propionic acid hcl salt
(3R)-3-amino-3-phenylpropanoic acid;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only