accessibility menu, dialog, popup

UserName

CAS RN 83649-48-3

NH2(R)HOO

CAS RN 83649-48-3

IUPAC Name: (3R)-3-amino-3-phenylpropanoic acid
Synonyms: (3R)-3-amino-3-phenylpropanoic acid
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: UJOYFRCOTPUKAK-MRVPVSSYSA-N
SMILES: N[C@H](CC(O)=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)