missing translation for 'onlineSavingsMsg'
Learn More

(R)-3,3,3-Trifluoro-2-hydroxy-2-methylpropionic Acid 98.0+%, TCI America™

Catalog No. T19001G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19001G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T19001G Supplier TCI America Supplier No. T19001G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 44864-47-3
Molecular Formula C4H5F3O3
Molecular Weight (g/mol) 158.08
MDL Number MFCD03095400
InChI Key CTGJACFEVDCYMC-UHFFFAOYNA-N
Synonym (R)-2-Hydroxy-2-(trifluoromethyl)propionic Acid, (R)-3,3,3-Trifluoro-2-hydroxyisobutyric Acid
PubChem CID 2760712
IUPAC Name 3,3,3-trifluoro-2-hydroxy-2-methylpropanoic acid
SMILES CC(O)(C(O)=O)C(F)(F)F

Specifications

Melting Point 110°C
Color White
Quantity 1 g
UN Number 3261
Formula Weight 158.08
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (R)-3,3,3-Trifluoro-2-hydroxy-2-methylpropionic Acid
TSCA No
Recommended Storage Refrigerator