accessibility menu, dialog, popup

UserName

CAS RN 44864-47-3

IUPAC Name: 3,3,3-trifluoro-2-hydroxy-2-methylpropanoic acid
Synonyms: 3,3,3-trifluoro-2-hydroxy-2-methylpropanoic acid
Molecular Weight (g/mol): 158.08
Molecular Formula: C4H5F3O3
InChi Key: CTGJACFEVDCYMC-UHFFFAOYNA-N
SMILES: CC(O)(C(O)=O)C(F)(F)F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results