missing translation for 'onlineSavingsMsg'
Learn More

(R)-2-Methylpyrrolidine p-toluenesulfonate, 97%, Thermo Scientific™

Catalog No. p-7034713
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5241003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5241003 Supplier Thermo Scientific Chemicals Supplier No. H5241003
Only null left

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 204387-55-3
Molecular Formula C12H19NO3S
Molecular Weight (g/mol) 257.348
MDL Number MFCD07368911
InChI Key ZXYDRZKLQBDQTN-QDXATWJZSA-N
Synonym r-2-methylpyrrolidine tosylate, r-2-methylpyrrolidine 4-methylbenzenesulfonate, r-2-methyl-pyrrolidine tosylate, 2r-2-methylpyrrolidine; para-toluene sulfonate, 2r-2-methylpyrrolidine tosylate, r-2-methylpyrrolidine p-toluenesulfonate, 2r-2-methylpyrrolidine toluene-4-sulphonate, 2r-2-methylpyrrolidine para-toluene sulfonate, r-2-methylpyrrolidine 4-methylphenylsulphonate, r-+-2-methylpyrrolidine p-toluenesulfonic acid salt
PubChem CID 20706077
IUPAC Name 4-methylbenzenesulfonic acid;(2R)-2-methylpyrrolidine
SMILES CC1CCCN1.CC1=CC=C(C=C1)S(=O)(=O)O

Specifications

Quantity 1 g
Formula Weight 257.35
Percent Purity 97%
Chemical Name or Material (R)-2-Methylpyrrolidine p-toluenesulfonate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures