missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-2-(Methoxymethyl)-1-pyrrolidinecarboxaldehyde 98.0+%, TCI America™

Catalog No. m11701g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M11701G Supplier TCI America Supplier No. M11701G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 121817-71-8
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.186
MDL Number MFCD00191670
InChI Key JNIOQRWRORXADR-SSDOTTSWSA-N
Synonym (R)-(+)-1-Formyl-2-(methoxymethyl)pyrrolidine
PubChem CID 12349568
IUPAC Name (2R)-2-(methoxymethyl)pyrrolidine-1-carbaldehyde
SMILES COCC1CCCN1C=O

Specifications

Color Yellow
Quantity 1 g
Formula Weight 143.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-(+)-2-(Methoxymethyl)-1-pyrrolidinecarboxaldehyde