accessibility menu, dialog, popup

UserName

CH3ONO

CAS RN 121817-71-8

IUPAC Name: 2-(methoxymethyl)pyrrolidine-1-carbaldehyde
Synonyms: 2-(methoxymethyl)pyrrolidine-1-carbaldehyde
Molecular Weight (g/mol): 143.19
Molecular Formula: C7H13NO2
InChi Key: JNIOQRWRORXADR-UHFFFAOYNA-N
SMILES: COCC1CCCN1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results