missing translation for 'onlineSavingsMsg'
Learn More

(R)-2-(Fmoc-amino)butyric acid, 98%

Catalog No. AAH6392314
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6392314 25 g
AAH6392303 1 g
AAH6392306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6392314 Supplier Thermo Scientific Chemicals Supplier No. H6392314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 170642-27-0 | C19H19NO4 | 325.364 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 170642-27-0
Molecular Formula C19H19NO4
Molecular Weight (g/mol) 325.364
MDL Number MFCD00270336
InChI Key XQIRYUNKLVPVRR-QGZVFWFLSA-N
Synonym fmoc-d-abu-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino butanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino butanoic acid, butanoic acid, 2-9h-fluoren-9-ylmethoxy carbonyl amino-, 2r, l-2-fmoc-amino butyric acid, fmoc-d-2-aminobutyric acid, fmoc-d-abu 2-oh, ambotzfaa1611, fmoc-d-2abu-oh
PubChem CID 853017
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)butanoic acid
SMILES CCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Quantity 25 g
Formula Weight 325.36
Percent Purity 98%
Physical Form Solid
Chemical Name or Material (R)-2-(Fmoc-amino)butyric acid
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only