missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-2-Diphenylphosphino-2'-methoxy-1,1'-binaphthyl 98.0+%, TCI America™

Catalog No. D27741G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27741G 1 g
D2774100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D27741G Supplier TCI America Supplier No. D27741G

Chemical Identifiers

CAS 145964-33-6
Molecular Formula C33H25OP
Molecular Weight (g/mol) 468.536
MDL Number MFCD00269686
InChI Key KRWTWSSMURUMDE-UHFFFAOYSA-N
Synonym (R)-(+)-MOP
PubChem CID 4190002
IUPAC Name [1-(2-methoxynaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane
SMILES COC1=C(C2=CC=CC=C2C=C1)C3=C(C=CC4=CC=CC=C43)P(C5=CC=CC=C5)C6=CC=CC=C6

Specifications

Melting Point 193°C
Color White-Yellow
Quantity 1 g
Formula Weight 468.54
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(+)-2-Diphenylphosphino-2′-methoxy-1,1′-binaphthyl