missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-2-Chloropropionic Acid 98.0+%, TCI America™

Catalog No. p-7125108
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16551G 1 g
C16555G 5 g
C165525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C16551G Supplier TCI America Supplier No. C16551G
Only null left

Chemical Identifiers

CAS 7474-05-7
Molecular Formula C3H4ClO2
Molecular Weight (g/mol) 107.51
MDL Number MFCD00064204
InChI Key GAWAYYRQGQZKCR-UWTATZPHSA-M
Synonym r-+-2-chloropropionic acid, r-2-chloropropanoic acid, r-2-chloropropionic acid, r-2-chloro-propionic acid, r +-2-chloropropionic acid, unii-f60st8319r, 2-chloropropionic acid, +, propanoic acid, 2-chloro-, 2r, d-2-chloropropionic acid, +-2-chloropropanoic acid
PubChem CID 2724540
IUPAC Name (2R)-2-chloropropanoic acid
SMILES CC(C(=O)O)Cl

Specifications

Color Colorless
Quantity 1 g
UN Number 2511
Formula Weight 108.52
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (R)-(+)-2-Chloropropionic Acid
EINECSNumber (2)-1157
TSCA No