accessibility menu, dialog, popup

UserName

CAS RN 7474-05-7

H3C(R)ClOO

CAS RN 7474-05-7

IUPAC Name: (2R)-2-chloropropanoate
Synonyms: (2R)-2-chloropropanoate
Molecular Weight (g/mol): 107.51
Molecular Formula: C3H4ClO2
InChi Key: GAWAYYRQGQZKCR-UWTATZPHSA-M
SMILES: C[C@@H](Cl)C([O-])=O
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results