missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Butanol 99.0+%, TCI America™

Catalog No. p-7118748
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B09261ML 1 mL
B09265ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B09261ML Supplier TCI America Supplier No. B09261ML
Only null left

Chemical Identifiers

CAS 14898-79-4
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00064280
InChI Key BTANRVKWQNVYAZ-SCSAIBSYSA-N
Synonym r---2-butanol, r-2-butanol, --2-butanol, 2r-butan-2-ol, r-butan-2-ol, r---sec-butanol, 2-butanol, 2r, unii-dlh38k423j, r---sec-butyl alcohol
PubChem CID 84682
ChEBI CHEBI:35686
IUPAC Name (2R)-butan-2-ol
SMILES CCC(C)O

Specifications

Color Colorless
Boiling Point 100°C
Quantity 1 mL
UN Number 1120
Formula Weight 74.12
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-(-)-2-Butanol
EINECSNumber (2)-3049
TSCA No