missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-2-Bromopropionic Acid 98.0+%, TCI America™

Supplier:  TCI America B175625G

 View more versions of this product

Catalog No. B175625G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

(R)-(+)-2-Bromopropionic Acid
104°C
MFCD00145205
3265
MONMFXREYOKQTI-UWTATZPHSA-N
(2R)-2-bromopropanoic acid
6992152
≥98.0% (GC)
10009-70-8
C3H5BrO2
25 g
r-+-2-bromopropionic acid, r-2-bromopropanoic acid, 2r-2-bromopropanoic acid, propanoic acid,2-bromo-, 2r, propanoic acid, 2-bromo-, 2r, r-bromopropionic acid, r-2-bromopropionic acid, r-2-bromo-propionic acid, 2r-2-bromo-propionic acid, r-+-2-bromopropionicacid
CC(C(=O)O)Br
152.975
152.98
Liquid

Chemical Identifiers

10009-70-8
152.975
MONMFXREYOKQTI-UWTATZPHSA-N
(2R)-2-bromopropanoic acid
C3H5BrO2
MFCD00145205
6992152
CC(C(=O)O)Br
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1167

TSCA : No

SDS
Documents

Documents

Promotions

Promotions