missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-2-Bromo-3-methylbutyric Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 44°C |
| Color | White-Yellow |
| Boiling Point | 100°C |
| Quantity | 5 g |
| UN Number | 3261 |
| Formula Weight | 181.03 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | (R)-2-Bromo-3-methylbutyric Acid |
Safety and Handling
| TSCA | No |