accessibility menu, dialog, popup

UserName

CAS RN 76792-22-8

CH3H3C(R)BrOHO

CAS RN 76792-22-8

IUPAC Name: (2R)-2-bromo-3-methylbutanoic acid
Synonyms: (2R)-2-bromo-3-methylbutanoic acid
Molecular Weight (g/mol): 181.03
Molecular Formula: C5H9BrO2
InChi Key: UEBARDWJXBGYEJ-SCSAIBSYSA-N
SMILES: CC(C)[C@@H](Br)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results