missing translation for 'onlineSavingsMsg'
Learn More

(R)-2-(Boc-amino)butyric acid, 95%

CAS: 45121-22-0 | C9H17NO4 | 203.238 g/mol

Supplier:  Thermo Scientific Chemicals H6399106

 View more versions of this product

Catalog No. AAH6399106


Only null left
Explore available promotions

Description

Description

(R)-2-(Boc-amino)butyric acid is used in coupling reaction. Also used in preparation of dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-2-(Boc-amino)butyric acid is used in coupling reaction. Also used in preparation of dyes.

Solubility
Soluble in DMF (1mmol in 1ml DMF).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
Specifications

Specifications

(R)-2-(Boc-amino)butyric acid
Yellow
MFCD00270335
boc-d-abu-oh, r-2-tert-butoxycarbonyl amino butanoic acid, r-n-boc-2-aminobutyric acid, boc-d-alpha-aminobutyric acid, boc-r-2-aminobutyric acid, boc-d-2-aminobutyric acid, 2r-2-tert-butoxycarbonyl amino butanoic acid, 2r-2-tert-butoxy carbonyl amino butanoic acid, butanoic acid, 2-1,1-dimethylethoxy carbonyl amino-, 2r, 2-r-tert-butoxycarbonylaminobutyric acid
PNFVIPIQXAIUAY-ZCFIWIBFSA-N
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
2755936
95%
45121-22-0
C9H17NO4
5 g
Soluble in DMF (1mmol in 1ml DMF).
CCC(C(=O)O)NC(=O)OC(C)(C)C
203.238
203.24
Liquid

Chemical Identifiers

45121-22-0
203.238
PNFVIPIQXAIUAY-ZCFIWIBFSA-N
2755936
CCC(C(=O)O)NC(=O)OC(C)(C)C
C9H17NO4
MFCD00270335
boc-d-abu-oh, r-2-tert-butoxycarbonyl amino butanoic acid, r-n-boc-2-aminobutyric acid, boc-d-alpha-aminobutyric acid, boc-r-2-aminobutyric acid, boc-d-2-aminobutyric acid, 2r-2-tert-butoxycarbonyl amino butanoic acid, 2r-2-tert-butoxy carbonyl amino butanoic acid, butanoic acid, 2-1,1-dimethylethoxy carbonyl amino-, 2r, 2-r-tert-butoxycarbonylaminobutyric acid
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only