CAS RN 45121-22-0
CAS RN 45121-22-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(R)-2-(Boc-amino)butyric acid, 95%
CAS: 45121-22-0 Molecular Formula: C9H17NO4 Molecular Weight (g/mol): 203.238 MDL Number: MFCD00270335 InChI Key: PNFVIPIQXAIUAY-ZCFIWIBFSA-N Synonym: boc-d-abu-oh,r-2-tert-butoxycarbonyl amino butanoic acid,r-n-boc-2-aminobutyric acid,boc-d-alpha-aminobutyric acid,boc-r-2-aminobutyric acid,boc-d-2-aminobutyric acid,2r-2-tert-butoxycarbonyl amino butanoic acid,2r-2-tert-butoxy carbonyl amino butanoic acid,butanoic acid, 2-1,1-dimethylethoxy carbonyl amino-, 2r,2-r-tert-butoxycarbonylaminobutyric acid PubChem CID: 2755936 IUPAC Name: (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid SMILES: CCC(C(=O)O)NC(=O)OC(C)(C)C