missing translation for 'onlineSavingsMsg'
Learn More

(R)-2-(Boc-amino)butyric acid, 95%

Catalog No. AAH6399103
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6399103 1 g
AAH6399106 5 g
AAH6399114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6399103 Supplier Thermo Scientific Chemicals Supplier No. H6399103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 45121-22-0 | C9H17NO4 | 203.238 g/mol

(R)-2-(Boc-amino)butyric acid is used in coupling reaction. Also used in preparation of dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-2-(Boc-amino)butyric acid is used in coupling reaction. Also used in preparation of dyes.

Solubility
Soluble in DMF (1mmol in 1ml DMF).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.

Chemical Identifiers

CAS 45121-22-0
Molecular Formula C9H17NO4
Molecular Weight (g/mol) 203.238
MDL Number MFCD00270335
InChI Key PNFVIPIQXAIUAY-ZCFIWIBFSA-N
Synonym boc-d-abu-oh, r-2-tert-butoxycarbonyl amino butanoic acid, r-n-boc-2-aminobutyric acid, boc-d-alpha-aminobutyric acid, boc-r-2-aminobutyric acid, boc-d-2-aminobutyric acid, 2r-2-tert-butoxycarbonyl amino butanoic acid, 2r-2-tert-butoxy carbonyl amino butanoic acid, butanoic acid, 2-1,1-dimethylethoxy carbonyl amino-, 2r, 2-r-tert-butoxycarbonylaminobutyric acid
PubChem CID 2755936
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES CCC(C(=O)O)NC(=O)OC(C)(C)C

Specifications

Color Yellow
Quantity 1 g
Solubility Information Soluble in DMF (1mmol in 1ml DMF).
Formula Weight 203.24
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material (R)-2-(Boc-amino)butyric acid
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only