missing translation for 'onlineSavingsMsg'
Learn More

(R)-2-(Boc-amino)-4-phenylbutyric acid, 98%

CAS: 82732-07-8 | C15H21NO4 | 279.336 g/mol

$88.44 - $388.23

Chemical Identifiers

CAS 82732-07-8
Molecular Formula C15H21NO4
Molecular Weight (g/mol) 279.336
MDL Number MFCD00076905
InChI Key MCODLPJUFHPVQP-GFCCVEGCSA-N
Synonym boc-d-homophenylalanine, boc-d-homophe-oh, boc-d-hophe-oh, boc-d-homo-phe, boc-d-hph-oh, n-boc-d-homophe-oh, boc-d-hph, boc-d-hphe-oh, boc-d-hfe-oh, 2r-2-tert-butoxycarbonyl amino-4-phenylbutanoic acid
PubChem CID 2755954
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-phenylbutanoic acid
SMILES CC(C)(C)OC(=O)NC(CCC1=CC=CC=C1)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6118303
View Documents
Thermo Scientific Chemicals
H6118303
1 g
Each for $88.44
Only null left
 
AAH6118306
View Documents
Thermo Scientific Chemicals
H6118306
5 g
Each for $388.23
Only null left
 
Description

Description

(R)-2-(Boc-amino)-4-phenylbutyric acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-2-(Boc-amino)-4-phenylbutyric acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

82732-07-8
279.336
MCODLPJUFHPVQP-GFCCVEGCSA-N
2755954
CC(C)(C)OC(=O)NC(CCC1=CC=CC=C1)C(=O)O
C15H21NO4
MFCD00076905
boc-d-homophenylalanine, boc-d-homophe-oh, boc-d-hophe-oh, boc-d-homo-phe, boc-d-hph-oh, n-boc-d-homophe-oh, boc-d-hph, boc-d-hphe-oh, boc-d-hfe-oh, 2r-2-tert-butoxycarbonyl amino-4-phenylbutanoic acid
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-phenylbutanoic acid

Specifications

71°C to 78°C
White
MFCD00076905
boc-d-homophenylalanine, boc-d-homophe-oh, boc-d-hophe-oh, boc-d-homo-phe, boc-d-hph-oh, n-boc-d-homophe-oh, boc-d-hph, boc-d-hphe-oh, boc-d-hfe-oh, 2r-2-tert-butoxycarbonyl amino-4-phenylbutanoic acid
MCODLPJUFHPVQP-GFCCVEGCSA-N
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-phenylbutanoic acid
279.336
279.33
Powder
82732-07-8
C15H21NO4
3653505
Slightly soluble in water.
CC(C)(C)OC(=O)NC(CCC1=CC=CC=C1)C(=O)O
1 g
2755954
98%
(R)-2-(Boc-amino)-4-phenylbutyric acid
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only