missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-2-Benzyloxypropionic acid, 98%

Catalog No. p-7058473
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1720506 5 g
AAL1720514 25 g
AAL1720522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1720506 Supplier Thermo Scientific Chemicals Supplier No. L1720506

CAS: 100836-85-9 | C10H12O3 | 180.203 g/mol

(R)-(+)-2-Benzyloxypropionic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(+)-2-Benzyloxypropionic acid is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 100836-85-9
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD06799065
InChI Key XWAVPOFYNPXXEL-MRVPVSSYSA-N
Synonym r-2-benzyloxy propanoic acid, r-+-2-benzyloxy propionic acid, r-+-2-benzyloxypropionic acid, o-benzyl-d-lactic acid, 2r-2-benzyloxy propanoic acid, propanoic acid, 2-phenylmethoxy-, 2r, r-benzyloxypropionic acid, r-2-benzyloxy-propionic acid, 2r-2-benzyloxy propionic acid, 2r-2-phenylmethoxypropanoic acid
PubChem CID 7000104
IUPAC Name (2R)-2-phenylmethoxypropanoic acid
SMILES CC(C(=O)O)OCC1=CC=CC=C1

Specifications

Melting Point 51°C to 53°C
Quantity 5 g
Beilstein 4675524
Solubility Information Insoluble in water.
Optical Rotation +90° (c=1 in IMS)
Formula Weight 180.2
Percent Purity 98%
Chemical Name or Material (R)-(+)-2-Benzyloxypropionic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only