missing translation for 'onlineSavingsMsg'
Learn More

(R)-2,6-Dichloro-3-fluoro-alpha-methylbenzyl Alcohol 98.0+%, TCI America™

Catalog No. p-7128347
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45771G 1 g
D45775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45771G Supplier TCI America Supplier No. D45771G

Chemical Identifiers

CAS 330156-50-8
Molecular Formula C8H7Cl2FO
Molecular Weight (g/mol) 209.041
MDL Number MFCD09863794
InChI Key JAOYKRSASYNDGH-SCSAIBSYSA-N
PubChem CID 11344814
IUPAC Name (1R)-1-(2,6-dichloro-3-fluorophenyl)ethanol
SMILES CC(C1=C(C=CC(=C1Cl)F)Cl)O

Specifications

Melting Point 43°C
Color White
Quantity 1 g
Formula Weight 209.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-2,6-Dichloro-3-fluoro-alpha-methylbenzyl Alcohol