accessibility menu, dialog, popup

UserName

CAS RN 330156-50-8

IUPAC Name: 1-(2,6-dichloro-3-fluorophenyl)ethan-1-ol
Synonyms: 1-(2,6-dichloro-3-fluorophenyl)ethanol
Molecular Weight (g/mol): 209.04
Molecular Formula: C8H7Cl2FO
InChi Key: JAOYKRSASYNDGH-UHFFFAOYNA-N
SMILES: CC(O)C1=C(Cl)C=CC(F)=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results