missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-2,2-Dimethyl-1,3-dioxolane-4-carboxaldehyde, 97%

Catalog No. AC369260010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC369260010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC369260010 Supplier Thermo Scientific Chemicals Supplier No. 369260010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 15186-48-8 | C6H10O3 | 130.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15186-48-8
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
MDL Number MFCD00269682
InChI Key YSGPYVWACGYQDJ-YFKPBYRVSA-N
Synonym r-+-2,2-dimethyl-1,3-dioxolane-4-carboxaldehyde, 4r-2,2-dimethyl-1,3-dioxolane-4-carbaldehyde, r-2,2-dimethyl-1,3-dioxolane-4-carbaldehyde, r-+-glyceraldehyde acetonide, 1,3-dioxolane-4-carboxaldehyde, 2,2-dimethyl-, r, acetone d-glyceraldehyde, +-2,2-dimethyl-1,3-dioxolane-4-carboxaldehyde, 1,3-dioxolane-4-carboxaldehyde, 2,2-dimethyl-, 4r, 4r-2,2-dimethyl-1,3-dioxolan-4-carbaldehyd, pubchem5767
PubChem CID 259712
IUPAC Name (4R)-2,2-dimethyl-1,3-dioxolane-4-carbaldehyde
SMILES CC1(OCC(O1)C=O)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 1.0450g/mL
Boiling Point 139.0°C
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Gravity 1.045
Specific Rotation +39° to +69° (20°C, 589nm) (C=2, chloroform)
Formula Weight 130.14
Percent Purity 97%
Concentration or Composition (by Analyte or Components) Contains various amounts of polymer
Physical Form Liquid
Chemical Name or Material (R)-(+)-2, 2-Dimethyl-1, 3-dioxolane-4-carboxaldehyde
Show More Show Less
Recommended Storage Crack to monomer by distillation before use

RUO – Research Use Only