missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 98%

CAS: 76189-55-4 | C44H32P2 | 622.69 g/mol

Supplier:  Thermo Scientific Chemicals B2378506

 View more versions of this product

Catalog No. AAB2378506


Only null left
Explore available promotions

Description

Description

(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthy is used as a chiral ligand for metal mediated asymmetric catalysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(+)-2,2′-Bis(diphenylphosphino)-1,1′-binaphthy is used as a chiral ligand for metal mediated asymmetric catalysis.

Solubility
Insoluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents.
Specifications

Specifications

(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
239°C to 241°C
MFCD00010805
4914063
binap, r-+-2,2'-bis diphenylphosphino-1,1'-binaphthyl, 2,2'-bis diphenylphosphino-1,1'-binaphthyl, s-binap, s---binap, s---2,2'-bis diphenylphosphino-1,1'-binaphthyl, r-+-binap, r-binap, rac-binap, 2,2'-bis diphenylphosphino-1,1'-binaphthalene
+230° (c=0.3 in Toluene)
[2'-(diphenylphosphanyl)-[1,1'-binaphthalen]-2-yl]diphenylphosphane
634876
98%
76189-55-4
C44H32P2
5 g
14,1223
MUALRAIOVNYAIW-UHFFFAOYSA-N
C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1C1=C(C=CC2=CC=CC=C12)P(C1=CC=CC=C1)C1=CC=CC=C1
622.69
622.69

Chemical Identifiers

76189-55-4
622.69
MUALRAIOVNYAIW-UHFFFAOYSA-N
634876
C44H32P2
MFCD00010805
binap, r-+-2,2'-bis diphenylphosphino-1,1'-binaphthyl, 2,2'-bis diphenylphosphino-1,1'-binaphthyl, s-binap, s---binap, s---2,2'-bis diphenylphosphino-1,1'-binaphthyl, r-+-binap, r-binap, rac-binap, 2,2'-bis diphenylphosphino-1,1'-binaphthalene
C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1C1=C(C=CC2=CC=CC=C12)P(C1=CC=CC=C1)C1=CC=CC=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only