Learn More
(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 98%
CAS: 76189-55-4 | C44H32P2 | 622.69 g/mol
Supplier: Thermo Scientific Chemicals B2378503
Description
(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthy is used as a chiral ligand for metal mediated asymmetric catalysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(R)-(+)-2,2′-Bis(diphenylphosphino)-1,1′-binaphthy is used as a chiral ligand for metal mediated asymmetric catalysis.
Solubility
Insoluble in water.
Notes
Air sensitive. Incompatible with oxidizing agents.
Specifications
| (R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl | |
| 239°C to 241°C | |
| MFCD00010805 | |
| 4914063 | |
| 14,1223 | |
| Insoluble in water. | |
| +230° (c=0.3 in Toluene) | |
| [1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane | |
| 634876 | |
| 98% |
| 76189-55-4 | |
| C44H32P2 | |
| 1 g | |
| Air Sensitive | |
| binap, r-+-2,2'-bis diphenylphosphino-1,1'-binaphthyl, 2,2'-bis diphenylphosphino-1,1'-binaphthyl, s-binap, s---binap, s---2,2'-bis diphenylphosphino-1,1'-binaphthyl, r-+-binap, r-binap, rac-binap, 2,2'-bis diphenylphosphino-1,1'-binaphthalene | |
| MUALRAIOVNYAIW-UHFFFAOYSA-N | |
| C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1C1=C(C=CC2=CC=CC=C12)P(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 622.69 | |
| 622.69 |
Chemical Identifiers
| 76189-55-4 | |
| 622.69 | |
| MUALRAIOVNYAIW-UHFFFAOYSA-N | |
| 634876 |
| C44H32P2 | |
| MFCD00010805 | |
| binap, r-+-2,2'-bis diphenylphosphino-1,1'-binaphthyl, 2,2'-bis diphenylphosphino-1,1'-binaphthyl, s-binap, s---binap, s---2,2'-bis diphenylphosphino-1,1'-binaphthyl, r-+-binap, r-binap, rac-binap, 2,2'-bis diphenylphosphino-1,1'-binaphthalene | |
| C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1C1=C(C=CC2=CC=CC=C12)P(C1=CC=CC=C1)C1=CC=CC=C1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only