missing translation for 'onlineSavingsMsg'
Learn More

(R)-2,2'-Bis[bis(3,5-trifluoromethylphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl, 97+%, Thermo Scientific Chemicals

Catalog No. AAH60561MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH60561MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH60561MD Supplier Thermo Scientific Chemicals Supplier No. H60561MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1365531-84-5 | Molecular Formula: C48H28F24O4P2 | Molecular Weight: 1186.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Insoluble in water.

Notes
Air sensitive. Store at room temperature.

Chemical Identifiers

CAS 1365531-84-5
Molecular Formula C48H28F24O4P2
Molecular Weight (g/mol) 1186.66
MDL Number MFCD19443625
InChI Key JDKXXRZDVSJSNQ-UHFFFAOYSA-N
Synonym r-2,2'-bis bis 3,5-trifluoromethylphenyl phosphino-4,4',6,6'-tetramethoxybiphenyl, s-2,2'-bis bis 3,5-trifluoromethylphenyl phosphino-4,4',6,6'-tetramethoxybiphenyl, ar-2,2',4,4'-tetramethoxy-6,6'-bis bis 3,5-bis trifluoromethyl phenyl phosphino biphenyl, r-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis bis 3,5-bis trifluoromethyl phenyl phosphine, r-2,2'-bis bis 3,5-trifluoromethylphenyl phosphino-4,4',6,6'-tetramethoxy-1,1'-biphenyl, s-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis bis 3,5-bis trifluoromethyl phenyl phosphine, s-2,2'-bis bis 3,5-trifluoromethylphenyl phosphino-4,4',6,6'-tetramethoxy-1,1'-biphenyl, bis 3,5-bis trifluoromethyl phenyl 2'-bis 3,5-bis trifluoromethyl phenyl phosphanyl-4,4',6,6'-tetramethoxy-1,1'-biphenyl-2-yl phosphane
PubChem CID 87111592
IUPAC Name [2-[2-bis[3,5-bis(trifluoromethyl)phenyl]phosphanyl-4,6-dimethoxyphenyl]-3,5-dimethoxyphenyl]-bis[3,5-bis(trifluoromethyl)phenyl]phosphane
SMILES COC1=CC(OC)=C(C(=C1)P(C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)C1=C(OC)C=C(OC)C=C1P(C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Specifications

Quantity 250 mg
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 1186.64
Percent Purity ≥97%
Chemical Name or Material (R)-2,2'-Bis[bis(3,5-trifluoromethylphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only