missing translation for 'onlineSavingsMsg'
Learn More

(R)-2,2'-Bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl, 97+%

Catalog No. p-7040354
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH60112MD 250 mg
AAH6011203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH60112MD Supplier Thermo Scientific Chemicals Supplier No. H60112MD
Only null left

CAS: 1365531-98-1 | C76H108O8P2 | 1211.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Insoluble in water.

Notes
Incompatible with light and oxidizing agents. Store in a cool place.

Chemical Identifiers

CAS 1365531-98-1
Molecular Formula C76H108O8P2
Molecular Weight (g/mol) 1211.64
MDL Number MFCD20265636
InChI Key KACYLFSRRUJDSY-UHFFFAOYSA-N
Synonym r-dtbm-garphos, 4,4',6,6'-tetramethoxy-1,1'-biphenyl-2,2'-diyl bis bis 3,5-di-tert-butyl-4-methoxyphenyl phosphine, ar-2,2'-bis bis 3,5-di-tert-butyl-4-methoxyphenyl phosphino-4,4'-dimethoxy-6,6'-dimethoxybiphenyl, r-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis bis 3,5-di-tert-butyl-4-methoxyphenyl phosphine, r-2,2'-bis bis 4-methoxy-3,5-di-t-butylphenyl phosphino-4,4',6,6'-tetramethoxy-1,1'-biphenyl, r-2,2'-bis bis 4-methoxy-3,5-di-t-butylphenyl phosphino-4,4',6,6'-tetramethoxybiphenyl, s-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis bis 3,5-di-tert-butyl-4-methoxyphenyl phosphine, s-2,2'-bis bis 4-methoxy-3,5-di-t-butylphenyl phosphino-4,4',6,6'-tetramethoxy-1,1'-biphenyl, 2'-bis 3,5-di-tert-butyl-4-methoxyphenyl phosphanyl-4,4',6,6'-tetramethoxy-1,1'-biphenyl-2-yl bis 3,5-di-tert-butyl-4-methoxyphenyl phosphane
PubChem CID 87110675
IUPAC Name [2-[2-bis(3,5-ditert-butyl-4-methoxyphenyl)phosphanyl-4,6-dimethoxyphenyl]-3,5-dimethoxyphenyl]-bis(3,5-ditert-butyl-4-methoxyphenyl)phosphane
SMILES CC(C)(C)C1=CC(=CC(=C1OC)C(C)(C)C)P(C2=CC(=C(C(=C2)C(C)(C)C)OC)C(C)(C)C)C3=CC(=CC(=C3C4=C(C=C(C=C4P(C5=CC(=C(C(=C5)C(C)(C)C)OC)C(C)(C)C)C6=CC(=C(C(=C6)C(C)(C)C)OC)C(C)(C)C)OC)OC)OC)OC

Specifications

Quantity 250 mg
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 1211.61
Percent Purity ≥97%
Chemical Name or Material (R)-2,2'-Bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only