missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(-)-2,2,2-Trifluoro-1-(9-anthryl)ethanol 99.0+%, TCI America™
$201.32 - $201.32
Chemical Identifiers
| CAS | 53531-34-3 |
|---|---|
| Molecular Formula | C16H11F3O |
| Molecular Weight (g/mol) | 276.26 |
| MDL Number | MFCD00001260,MFCD00062967 |
| InChI Key | ICZHJFWIOPYQCA-UHFFFAOYNA-N |
| Synonym | r---2,2,2-trifluoro-1-9-anthryl ethanol, 1r-1-anthracen-9-yl-2,2,2-trifluoroethanol, r-1-anthracen-9-yl-2,2,2-trifluoroethanol, 1r-1-9-anthracenyl-2,2,2-trifluoroethanol, 1r-1-9-anthracenyl-2,2,2-trifluoroethanol, r-+-2,2,2-trifluoro-1-9-anthryl ethanol, r-+-2,2,2-trifluoro-1-9-anthryl ethanol, r---, 1r-1-anthracen-9-yl-2,2,2-tris fluoranyl ethanol, r---alpha-trifluoromethyl-9-anthracenemethanol, r---alpha-trifluoromethyl-9-anthracenemethanol |
| PubChem CID | 7006444 |
| IUPAC Name | 1-(anthracen-9-yl)-2,2,2-trifluoroethan-1-ol |
| SMILES | OC(C1=C2C=CC=CC2=CC2=CC=CC=C12)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T1520100MG
|
TCI America
T1520100MG |
100 mg |
Each for $201.32
|
|
|||||
Chemical Identifiers
| 53531-34-3 | |
| 276.26 | |
| ICZHJFWIOPYQCA-UHFFFAOYNA-N | |
| 7006444 | |
| OC(C1=C2C=CC=CC2=CC2=CC=CC=C12)C(F)(F)F |
| C16H11F3O | |
| MFCD00001260,MFCD00062967 | |
| r---2,2,2-trifluoro-1-9-anthryl ethanol, 1r-1-anthracen-9-yl-2,2,2-trifluoroethanol, r-1-anthracen-9-yl-2,2,2-trifluoroethanol, 1r-1-9-anthracenyl-2,2,2-trifluoroethanol, 1r-1-9-anthracenyl-2,2,2-trifluoroethanol, r-+-2,2,2-trifluoro-1-9-anthryl ethanol, r-+-2,2,2-trifluoro-1-9-anthryl ethanol, r---, 1r-1-anthracen-9-yl-2,2,2-tris fluoranyl ethanol, r---alpha-trifluoromethyl-9-anthracenemethanol, r---alpha-trifluoromethyl-9-anthracenemethanol | |
| 1-(anthracen-9-yl)-2,2,2-trifluoroethan-1-ol |
Specifications
| 53531-34-3 | |
| C16H11F3O | |
| 100 mg | |
| ICZHJFWIOPYQCA-UHFFFAOYNA-N | |
| 1-(anthracen-9-yl)-2,2,2-trifluoroethan-1-ol | |
| 7006444 | |
| ≥99.0% (GC) | |
| (R)-(-)-2,2,2-Trifluoro-1-(9-anthryl)ethanol [e.e. Determination Reagent by NMR] |
| 134°C | |
| MFCD00001260,MFCD00062967 | |
| r---2,2,2-trifluoro-1-9-anthryl ethanol, 1r-1-anthracen-9-yl-2,2,2-trifluoroethanol, r-1-anthracen-9-yl-2,2,2-trifluoroethanol, 1r-1-9-anthracenyl-2,2,2-trifluoroethanol, 1r-1-9-anthracenyl-2,2,2-trifluoroethanol, r-+-2,2,2-trifluoro-1-9-anthryl ethanol, r-+-2,2,2-trifluoro-1-9-anthryl ethanol, r---, 1r-1-anthracen-9-yl-2,2,2-tris fluoranyl ethanol, r---alpha-trifluoromethyl-9-anthracenemethanol, r---alpha-trifluoromethyl-9-anthracenemethanol | |
| OC(C1=C2C=CC=CC2=CC2=CC=CC=C12)C(F)(F)F | |
| 276.26 | |
| 276.26 | |
| Crystalline Powder |
Safety and Handling
TSCA : No