missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2,2,2-Trifluoro-1-(9-anthryl)ethanol 99.0+%, TCI America™

Catalog No. T1520100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1520100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T1520100MG Supplier TCI America Supplier No. T1520100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 53531-34-3
Molecular Formula C16H11F3O
Molecular Weight (g/mol) 276.26
MDL Number MFCD00001260,MFCD00062967
InChI Key ICZHJFWIOPYQCA-UHFFFAOYNA-N
Synonym r---2,2,2-trifluoro-1-9-anthryl ethanol, r---1-9-anthryl-2,2,2-trifluoroethanol, r-1-anthracen-9-yl-2,2,2-trifluoroethanol, 1r-1-9-anthracenyl-2,2,2-trifluoroethanol, 1r-1-9-anthryl-2,2,2-trifluoroethan-1-ol, r-+-2,2,2-trifluoro-1-9-anthryl ethanol, r---, a-trifluoromethyl anthracene-9-methanol, 1r-1-anthracen-9-yl-2,2,2-tris fluoranyl ethanol, r---alpha-trifluoromethyl-9-anthracenemethanol, 1r-1-anthracen-9-yl-2,2,2-trifluoroethanol
PubChem CID 7006444
IUPAC Name 1-(anthracen-9-yl)-2,2,2-trifluoroethan-1-ol
SMILES OC(C1=C2C=CC=CC2=CC2=CC=CC=C12)C(F)(F)F

Specifications

Melting Point 134°C
Quantity 100 mg
Formula Weight 276.26
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(-)-2,2,2-Trifluoro-1-(9-anthryl)ethanol [e.e. Determination Reagent by NMR]