missing translation for 'onlineSavingsMsg'
Learn More

(R)-2-(1-Hydroxyethyl)pyridine, 98%, Thermo Scientific Chemicals

$79.72 - $79.72

Chemical Identifiers

CAS 27911-63-3
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.15
MDL Number MFCD04972322
InChI Key PPHIIIRFJKDTLG-ZCFIWIBFSA-N
Synonym r-1-pyridin-2-yl ethanol, r-2-1-hydroxyethyl pyridine, r-1-2-pyridyl ethanol, 1r-1-pyridin-2-yl ethan-1-ol, r-alpha-methyl-2-pyridinemethanol, 1r-1-pyridin-2-ylethanol, 1r-1-pyridin-2-yl ethanol, pubchem5699, r-1-pyridin-2-yl-ethanol, 1r-1-2-pyridyl ethanol
PubChem CID 642847
IUPAC Name (1R)-1-pyridin-2-ylethanol
SMILES CC(C1=CC=CC=N1)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC382862500
SDS View Documents
Thermo Scientific Chemicals
382862500
250 mg Glass Bottle
Each for $79.72
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

27911-63-3
123.15
PPHIIIRFJKDTLG-ZCFIWIBFSA-N
642847
CC(C1=CC=CC=N1)O
C7H9NO
MFCD04972322
r-1-pyridin-2-yl ethanol, r-2-1-hydroxyethyl pyridine, r-1-2-pyridyl ethanol, 1r-1-pyridin-2-yl ethan-1-ol, r-alpha-methyl-2-pyridinemethanol, 1r-1-pyridin-2-ylethanol, 1r-1-pyridin-2-yl ethanol, pubchem5699, r-1-pyridin-2-yl-ethanol, 1r-1-2-pyridyl ethanol
(1R)-1-pyridin-2-ylethanol

Specifications

27911-63-3
100.0
Authentic
Glass Bottle
1.5270 to 1.529
250 mg
+ 24
PPHIIIRFJKDTLG-ZCFIWIBFSA-N
(1R)-1-pyridin-2-ylethanol
642847
98%
(R)-2-(1-Hydroxyethyl)pyridine
97.5
Pink
97.5% min. (GC)
C7H9NO
MFCD04972322
+24° (31°C c=1 toluene)
r-1-pyridin-2-yl ethanol, r-2-1-hydroxyethyl pyridine, r-1-2-pyridyl ethanol, 1r-1-pyridin-2-yl ethan-1-ol, r-alpha-methyl-2-pyridinemethanol, 1r-1-pyridin-2-ylethanol, 1r-1-pyridin-2-yl ethanol, pubchem5699, r-1-pyridin-2-yl-ethanol, 1r-1-2-pyridyl ethanol
CC(C1=CC=CC=N1)O
123.15
123.15
Crystals
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only