accessibility menu, dialog, popup

UserName

H3C(R)OHN

CAS RN 27911-63-3

IUPAC Name: (1R)-1-(pyridin-2-yl)ethan-1-ol
Synonyms: (1R)-1-(pyridin-2-yl)ethanol
Molecular Weight (g/mol): 123.16
Molecular Formula: C7H9NO
InChi Key: PPHIIIRFJKDTLG-ZCFIWIBFSA-N
SMILES: C[C@@H](O)C1=CC=CC=N1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results