missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1-Phenylpropylamine, ChiPros 99+%, ee 98%

Catalog No. AAL1631914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1631914 25 g
AAL1631906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1631914 Supplier Thermo Scientific Chemicals Supplier No. L1631914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3082-64-2 | C9H13N | 135.21 g/mol

For synthesis of optically active products

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3082-64-2
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00083057
InChI Key AQFLVLHRZFLDDV-SECBINFHSA-N
Synonym r-+-1-phenylpropylamine, r-1-phenylpropan-1-amine, 1r-1-phenylpropan-1-amine, r-1-phenylpropylamine, r-+-alpha-ethylbenzylamine, 1r-1-phenylpropylamine, r-+-methylbenzylamine, r-+-1-amino-1-phenylpropane, r +-a-ethylbenzylamine, r-+-, a-ethylbenzylamine
PubChem CID 5324978
IUPAC Name (1R)-1-phenylpropan-1-amine
SMILES CCC(C1=CC=CC=C1)N

Specifications

Melting Point -69°C
Density 0.93
Boiling Point 205°C
Flash Point 77°C (170°F)
Odor Amine-like
Refractive Index 1.52
Quantity 25 g
UN Number UN2735
Beilstein 3029904
Sensitivity Air Sensitive
Optical Rotation +20° (Neat)
Formula Weight 135.21
Percent Purity ≥99%
Assay ee 98%
Chemical Name or Material (R)-(+)-1-Phenylpropylamine
Show More Show Less
Hazard Category H227-H302-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 608-544-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only