missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1-Phenyl-1-propanol 98.0+%, TCI America™

Catalog No. P19301G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P19301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P19301G Supplier TCI America Supplier No. P19301G

Chemical Identifiers

CAS 1565-74-8
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00064279
InChI Key DYUQAZSOFZSPHD-SECBINFHSA-N
Synonym r-+-1-phenyl-1-propanol, r-1-phenylpropan-1-ol, 1r-1-phenylpropan-1-ol, r-1-phenyl-1-propanol, r-1-phenyl-propanol, r-+-alpha-ethylbenzyl alcohol, r-+-1-phenylpropanol, r-1-phenylpropanol, 1-phenyl-1-propanol #, r-1-phenyl-1propanol
PubChem CID 640199
IUPAC Name (1R)-1-phenylpropan-1-ol
SMILES CCC(C1=CC=CC=C1)O

Specifications

Color Yellow
Boiling Point 105°C
Quantity 1 g
Formula Weight 136.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-(+)-1-Phenyl-1-propanol
EINECSNumber (3)-1049&(3)-1034
RTECSNumber DO5470000
TSCA No