missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1-Phenyl-1-propanol, 99%

Catalog No. AAL05681ME
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL05681ME 500 mg
AAL05681MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL05681ME Supplier Thermo Scientific Chemicals Supplier No. L05681ME
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1565-74-8 | C9H12O | 136.194 g/mol

For synthesis of optically active products

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1565-74-8
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00064279
InChI Key DYUQAZSOFZSPHD-SECBINFHSA-N
Synonym r-+-1-phenyl-1-propanol, r-1-phenylpropan-1-ol, 1r-1-phenylpropan-1-ol, r-1-phenyl-1-propanol, r-1-phenyl-propanol, r-+-alpha-ethylbenzyl alcohol, r-+-1-phenylpropanol, r-1-phenylpropanol, 1-phenyl-1-propanol #, r-1-phenyl-1propanol
PubChem CID 640199
IUPAC Name (1R)-1-phenylpropan-1-ol
SMILES CCC(C1=CC=CC=C1)O

Specifications

Density 0.993
Boiling Point 217°C to 219°C
Flash Point 95°C (203°F)
Refractive Index 1.52
Quantity 500 mg
Beilstein 2041555
Optical Rotation +47° (c=2.2 in Hexane)
Formula Weight 136.19
Percent Purity 99%
Chemical Name or Material (R)-(+)-1-Phenyl-1-propanol
Hazard Category H227-H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P210-P264b-P270-P280-P301+P312-P330-P370+P378q-P501c
RTECSNumber DO5470000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only