missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1-Methoxy-2-propanol 98.0+%, TCI America™

Catalog No. M21665G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21665G 5 g
M216625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21665G Supplier TCI America Supplier No. M21665G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4984-22-9
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.122
MDL Number MFCD01632587
InChI Key ARXJGSRGQADJSQ-SCSAIBSYSA-N
Synonym r---1-methoxy-2-propanol, 2r-1-methoxypropan-2-ol, r-1-methoxypropan-2-ol, r-1-methoxy-2-propanol, r-1-methoxy-propan-2-ol, 2-propanol, 1-methoxy-, 2r, 20s-2,5,8,11,14,17-hexamethyl-3,6,9,12,15,18-hexaoxahenicosane-1,20-diol, pubchem6714, r-methoxy-propan-2-ol
PubChem CID 2733589
IUPAC Name (2R)-1-methoxypropan-2-ol
SMILES CC(COC)O

Specifications

Color Colorless
Boiling Point 120°C
Quantity 5 g
UN Number 3092
Formula Weight 90.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-(-)-1-Methoxy-2-propanol
EINECSNumber (2)-0404
RTECSNumber UB7700000
TSCA No