accessibility menu, dialog, popup

UserName

CAS RN 4984-22-9

IUPAC Name: 1-methoxypropan-2-ol
Synonyms: methoxyisopropanol
Molecular Weight (g/mol): 90.12
Molecular Formula: C4H10O2
InChi Key: ARXJGSRGQADJSQ-UHFFFAOYNA-N
SMILES: COCC(C)O
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)