missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1-Methoxy-2-propanol, 98+%

Catalog No. p-158247
Change view
Click to view available options
Quantity:
1 mL
10 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC331780010 1 mL Glass bottle
AC331780100 10 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC331780010 Supplier Thermo Scientific Chemicals Supplier No. 331780010
Only null left

CAS: 4984-22-9 | C4H10O2 | 90.122 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4984-22-9
MDL Number MFCD01632587
InChI Key ARXJGSRGQADJSQ-SCSAIBSYSA-N
Synonym r---1-methoxy-2-propanol, 2r-1-methoxypropan-2-ol, r-1-methoxypropan-2-ol, r-1-methoxy-2-propanol, r-1-methoxy-propan-2-ol, 2-propanol, 1-methoxy-, 2r, 20s-2,5,8,11,14,17-hexamethyl-3,6,9,12,15,18-hexaoxahenicosane-1,20-diol, pubchem6714, r-methoxy-propan-2-ol
PubChem CID 2733589
IUPAC Name (2R)-1-methoxypropan-2-ol
SMILES CC(COC)O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -97.0°C
Color Colorless
Density 0.9200g/mL
Boiling Point 120.0°C
Flash Point 30°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Molecular Formula C4H10O2
Refractive Index 1.4020 to 1.4040
Linear Formula CH3OCH2CH(OH)CH3
Quantity 1 mL
Specific Gravity 0.92
Specific Rotation Condition −22° (20°C c=10,CHCl3)
Specific Rotation − 22.00
Molecular Weight (g/mol) 90.122
Formula Weight 90.12
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material (R)-(-)-1-Methoxy-2-propanol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
TSCA TSCA

RUO – Research Use Only