missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1-Cyclohexylethylamine, ChiPros 98%, ee 94+%

Catalog No. AAL1905114
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1905114 25 g
AAL1905103 1 g
AAL1905106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1905114 Supplier Thermo Scientific Chemicals Supplier No. L1905114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5913-13-3 | C8H17N | 127.23 g/mol

(R)-(-)-1-Cyclohexylethylamine is used in the preparation of benzylidene(1-cyclohexylethyl)amine by reacting with benzaldehyde. It is also used to prepare chiral polymethacrylamides, which are employed as stationary phases for the liquid-chromatographic resolution of racemic amides. In addition to this, it is used in the preparation of polyamides bound to silica gel for high performance liquid chromatography, which are pressure-stable.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-1-Cyclohexylethylamine is used in the preparation of benzylidene(1-cyclohexylethyl)amine by reacting with benzaldehyde. It is also used to prepare chiral polymethacrylamides, which are employed as stationary phases for the liquid-chromatographic resolution of racemic amides. In addition to this, it is used in the preparation of polyamides bound to silica gel for high performance liquid chromatography, which are pressure-stable.

Solubility
Miscible with acetone, chloroform,dimethyl sulfoxide, ethanol, methanol, ether, petroleum ether and dichloromethane. Insoluble in water.

Notes
Hygroscopic. Air sensitive. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 5913-13-3
Molecular Formula C8H17N
Molecular Weight (g/mol) 127.23
MDL Number MFCD00043338
InChI Key XBWOPGDJMAJJDG-UHFFFAOYNA-N
Synonym r---1-cyclohexylethylamine, r-1-cyclohexylethylamine, 1r-1-cyclohexylethanamine, r-1-cyclohexylethanamine, 1r-1-cyclohexylethylamine, chembl19290, 1r-1-cyclohexylethan-1-amine, 1-cyclohexyl-ethylamine, 1-cyclohexylethanamine #, r---cyclohexylethylamine
PubChem CID 10997046
SMILES CC(N)C1CCCCC1

Specifications

Density 0.866
pH 11
Boiling Point 177°C to 178°C
Flash Point 51°C (123°F)
Odor Amine-like
Refractive Index 1.462
Quantity 25 g
UN Number UN2733
Beilstein 2935070
Sensitivity Air Sensitive
Solubility Information Miscible with acetone,chloroform,dimethyl sulfoxide,ethanol,methanol,ether,petroleum ether and dichloromethane. Insoluble in water.
Optical Rotation −4° (Neat)
IUPAC Name (1R)-1-cyclohexylethanamine
Formula Weight 127.23
Percent Purity 98%
Assay ee ≥94%
Chemical Name or Material (R)-(-)-1-Cyclohexylethylamine
Show More Show Less
Hazard Category H226-H314-H335
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 227-634-2
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only