accessibility menu, dialog, popup

UserName

CAS RN 5913-13-3

IUPAC Name: 1-cyclohexylethan-1-amine
Synonyms: 1-cyclohexylethanamine
Molecular Weight (g/mol): 127.23
Molecular Formula: C8H17N
InChi Key: XBWOPGDJMAJJDG-UHFFFAOYNA-N
SMILES: CC(N)C1CCCCC1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)