missing translation for 'onlineSavingsMsg'
Learn More

(R)-1-Carbobenzoxy-3-pyrrolidinol 98.0+%, TCI America™

Catalog No. p-7125543
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25851G 1 g
C25855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C25851G Supplier TCI America Supplier No. C25851G
Only null left

Chemical Identifiers

CAS 100858-33-1
Molecular Formula C12H15NO3
Molecular Weight (g/mol) 221.26
MDL Number MFCD07368258
InChI Key MBLJFGOKYTZKMH-UHFFFAOYNA-N
Synonym r---1-cbz-3-pyrrolidinol, benzyl 3r-3-hydroxypyrrolidine-1-carboxylate, r-1-cbz-3-pyrrolidinol, r-benzyl 3-hydroxypyrrolidine-1-carboxylate, r-n-cbz-3-hydroxypyrrolidine, r-1-carbobenzoxy-3-pyrrolidinol, r-1-cbzl-3-pyrrolidinol, r-n-z-3-pyrrolidinol, r---1-cbz-3-hydroxy-pyrrolidine, 3r-n-cbz-3-pyrrolidinol
PubChem CID 11183628
IUPAC Name benzyl 3-hydroxypyrrolidine-1-carboxylate
SMILES OC1CCN(C1)C(=O)OCC1=CC=CC=C1

Specifications

Melting Point 77°C
Color Yellow
Quantity 1 g
Formula Weight 221.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-1-Carbobenzoxy-3-pyrrolidinol